Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Naphthalene-2-boronic acid pinacol ester, 97%

Click to view available options
Quantity:
1g
5g
256652-04-7
C16H19BO2
254.136
MFCD05663868
SPPZBAGKKBHZRW-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-naphthalen-2-yl-1,3,2-dioxaborolane,naphthalene-2-boronic acid pinacol ester,naphthalene-2-boronic acid, pinacol ester,2-naphthaleneboronic acid, pinacol ester,2-naphth-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,4,4,5,5-tetramethyl-2-2-naphthyl-1,3,2-dioxaborolane,1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-naphthalenyl,2-naphthaleneboronic acid pinacol ester,4,4,5,5-tetramethyl-2-2-naphthalenyl-1,3,2-dioxaborolane,amtb668
10753409
4,4,5,5-tetramethyl-2-naphthalen-2-yl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC3=CC=CC=C3C=C2
256652-04-7
C16H19BO2
254.136
MFCD05663868
SPPZBAGKKBHZRW-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-naphthalen-2-yl-1,3,2-dioxaborolane,naphthalene-2-boronic acid pinacol ester,naphthalene-2-boronic acid, pinacol ester,2-naphthaleneboronic acid, pinacol ester,2-naphth-2-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,4,4,5,5-tetramethyl-2-2-naphthyl-1,3,2-dioxaborolane,1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-naphthalenyl,2-naphthaleneboronic acid pinacol ester,4,4,5,5-tetramethyl-2-2-naphthalenyl-1,3,2-dioxaborolane,amtb668
10753409
4,4,5,5-tetramethyl-2-naphthalen-2-yl-1,3,2-dioxaborolane
B1(OC(C(O1)(C)C)(C)C)C2=CC3=CC=CC=C3C=C2
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Quantity 5g
Formula Weight 254.14
Physical Form Solid
Percent Purity 97%
Chemical Name or Material Naphthalene-2-boronic acid pinacol ester