Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Naphthalene-2-sulfonyl chloride, 97%

Click to view available options
Quantity:
25g
100g
500g
93-11-8
C10H7ClO2S
226.674
MFCD00004087
OPECTNGATDYLSS-UHFFFAOYSA-N
2-naphthalenesulfonyl chloride,2-naphthylsulfonyl chloride,beta-naphthalenesulfonyl chloride,naphthalene-2-sulfonic acid chloride,.beta.-naphthalenesulfochloride,naphthalene-2-sulphonyl chloride,2-naphthalenesulfonylchloride,2-naphthalenylsulfonyl chloride,2-naphthalene sulfonyl chloride,.beta.-naphthalenesulfonyl chloride
7125
naphthalene-2-sulfonyl chloride
C1=CC=C2C=C(C=CC2=C1)S(=O)(=O)Cl
93-11-8
C10H7ClO2S
226.674
MFCD00004087
OPECTNGATDYLSS-UHFFFAOYSA-N
2-naphthalenesulfonyl chloride,2-naphthylsulfonyl chloride,beta-naphthalenesulfonyl chloride,naphthalene-2-sulfonic acid chloride,.beta.-naphthalenesulfochloride,naphthalene-2-sulphonyl chloride,2-naphthalenesulfonylchloride,2-naphthalenylsulfonyl chloride,2-naphthalene sulfonyl chloride,.beta.-naphthalenesulfonyl chloride
7125
naphthalene-2-sulfonyl chloride
C1=CC=C2C=C(C=CC2=C1)S(=O)(=O)Cl
Sensitivity Moisture sensitive
Quantity 500g
Melting Point 74°C to 78°C
Boiling Point 200°C to 202°C (13mmHg)
UN Number UN3261
Beilstein 641898
Formula Weight 226.68
Percent Purity 97%
Chemical Name or Material Naphthalene-2-sulfonyl chloride

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 202-219-9

TSCA : Yes

Recommended Storage : Ambient temperatures