Learn More
Naphthalene, 99%, Thermo Scientific Chemicals
Description
Naphthalene is formerly important in dyestuff manufacture. It acts as a precursor to phthalic anhydride. Naphthalene is used in the manufacturing of phthalic anhydride, naphthalene sulfonic acids and hydrogenated naphthalenes such as 1,2,3,4-tetrahydronaphthalene, which is used as a precursor for various dyestuffs, pigments, rubber processing chemicals and in pharmaceuticals. Its molten form is used as solvent replacing the high- boiling solvents like dichlorobenzene, nitrobenzene and durene. It is involved as reaction medium for in the Diels-Alder reaction between C60 with anthracene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 80°C to 82°C |
Density | 0.9628 g/mL |
Boiling Point | 218°C |
Flash Point | 78°C (172°F) |
Odor | Aromatic |
Refractive Index | 1.5821 |
Quantity | 50 g |
UN Number | UN1334 |
Beilstein | 1421310 |
Merck Index | 14,6370 |
Show More |
Safety and Handling
Hazard Category | H228-H302-H350 |
Hazard Statement | GHS H Statement H228-H351-H302 Flammable solid. Suspected of causing cancer. Harmful if swallowed. |
Precautionary Statement | P201-P202-P210-P240-P241-P260-P264b-P270-P271-P281-P301+P312-P304+P340-P308+P313-P330-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: NAPHTHALENE, REFINED |
EINECSNumber | 202-049-5 |
RTECSNumber | QJ0525000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.