Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naphthalene, 99%, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
2.5kg
5kg
Packaging:
Plastic bottle
Plastic drum
91-20-3
C10H8
128.17
MFCD00001742
UFWIBTONFRDIAS-UHFFFAOYSA-N
naphthalin,naphthene,camphor tar,tar camphor,white tar,albocarbon,naphthaline,moth flakes,moth balls,naphtalene
931
CHEBI:16482
naphthalene
C1=CC=C2C=CC=CC2=C1
91-20-3
C10H8
128.17
MFCD00001742
UFWIBTONFRDIAS-UHFFFAOYSA-N
naphthalin,naphthene,camphor tar,tar camphor,white tar,albocarbon,naphthaline,moth flakes,moth balls,naphtalene
931
CHEBI:16482
naphthalene
C1=CC=C2C=CC=CC2=C1
Specific Gravity 0.99
Quantity 2.5kg
Infrared Spectrum Authentic
Melting Point 79.0°C to 82.0°C
Color White
Boiling Point 218.0°C
Density 0.9900g/mL
Flash Point 78°C
Packaging Plastic bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula C10H8
Beilstein 05,IV,164
Merck Index 15,6455
Solubility Information Solubility in water: 30mg/l (25°C). Other solubilities: soluble in ethanol,,very soluble in acetone,chloroform,ether,,benzene,toluene,carbon disulfide
Formula Weight 128.17
Physical Form Crystals, Crystalline Flakes, Tablets, Balls or Crystalline Powder
Percent Purity 99%
Chemical Name or Material Naphthalene
Show More Show Less

GHS H Statement
Suspected of causing cancer.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
Flammable solid.

GHS P Statement
Obtain special instructions before use.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces.
- No smokin

GHS Signal Word: Warning

EINECSNumber : 202-049-5