Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naproxen sodium, Tocris Bioscience™

Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
26159-34-2
C14H13NaO3
252.25
MFCD00058507
CDBRNDSHEYLDJV-FVGYRXGTSA-M
naproxen sodium, anaprox, miranax, naprelan, naproxen sodium salt, anapran, anaprotab, aprowell, floginex, flogogin
23681059
CHEBI:7477
sodium (2S)-2-(6-methoxynaphthalen-2-yl)propanoate
[Na+].COC1=CC=C2C=C(C=CC2=C1)[C@H](C)C([O-])=O
This item is not returnable. View return policy
26159-34-2
C14H13NaO3
252.25
MFCD00058507
CDBRNDSHEYLDJV-FVGYRXGTSA-M
naproxen sodium, anaprox, miranax, naprelan, naproxen sodium salt, anapran, anaprotab, aprowell, floginex, flogogin
23681059
CHEBI:7477
sodium (2S)-2-(6-methoxynaphthalen-2-yl)propanoate
[Na+].COC1=CC=C2C=C(C=CC2=C1)[C@H](C)C([O-])=O

Cyclooxygenase inhibitor

Non-selective cyclooxygenase (COX) inhibitor that displays anti-inflammatory, antipyretic and analgesic effects. Has a neuroprotective role against colchicine-induced cognitive impairment and oxidative stress.
Quantity 50 mg
Formula Weight 252.24
Percent Purity >99%
Chemical Name or Material Naproxen sodium

Recommended Storage : Desiccate at Room Temperature