Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAPSA Mouse anti-Human, Clone: OTI1B5, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527023

Catalog No. PIMA527023


Add to cart

Description

Description

Napsin 1 is an aspartic protease that is generally expressed in the lung, but also present in kidney and in metastatic carcinomas throughout the body. Napsin 1 processes surfactant protein B in healthy pulmonary and renal tissues, but is abnormally regulated when carcinogenesis occurs. Thus, many researchers have found napsin evaluation to be useful in identifying several cancers, and differentiating primary tumors from metastases. When combined with thyroid transcription factor analysis, napsin can be used as a specific biomarker for differentiation of carcinoma types. Napsin has also been associated with pulmonary alveolar proteinosis as well as in protein catabolism in renal tubules.
Specifications

Specifications

NAPSA
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O96009
KAP, Kdap, NAP1, NAPA, SNAPA, ASP4, TA01/TA02, asp 4, aspartyl protease 4, kidney-derived aspartic protease-like protein, napsin-1, napsin-A, pronapsin A
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9476
Immunohistochemistry (Paraffin), Western Blot
OTI1B5
Unconjugated
NAPSA
Liquid
NAPSA
Human recombit protein fragment corresponding to amino acids 64-244 of NAPSA produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit