Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Napsin A Antibody (NAPSA/1238) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
$627.50
Specifications
Antigen | Napsin A |
---|---|
Clone | NAPSA/1238 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1 - 2 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB254383100
|
Novus Biologicals
NBP254383100UG |
100 μg |
Each for $627.50
|
|
|||||
NB254383200
|
Novus Biologicals
NBP254383200UG |
200 μg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Napsin A Monoclonal specifically detects Napsin A in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Protein Array, CyTOF-ready.Specifications
Napsin A | |
1 mg/mL | |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
Unconjugated | |
Mouse | |
Cancer, Cell Biology, Cellular Markers | |
PBS with No Preservative | |
9476 | |
Recombinant fragment of human Napsin-A protein | |
Primary | |
Store at -20 to -80C. Avoid freeze-thaw cycles. | |
37 kDa |
NAPSA/1238 | |
Western Blot 1 - 2 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Asp 4, ASP4, Aspartyl protease 4, EC 3.4.23, EC 3.4.23.-, EC 3.4.23.15, EC 3.4.23.3, EC 3.4.23.5, KAP, Kdap, NAP1napsin-A, NAPApronapsin A, NAPSA, napsin A aspartic peptidase, napsin-1, SNAPA, TA01/TA02 | |
NAPSA | |
IgG κ | |
Protein A purified | |
Napsin is a pepsin-like aspartic proteinase connected with maturation of surfactant protein B.There are two closely related napsins, napsin A and napsin B. Napsin A is expressed as a single chain protein. Immunohistochemical studies revealed high expression levels of napsin A in human lung and kidney but low expression in spleen. Napsin A is expressed in type II pneumocytes and in adenocarcinomas of lung. The high specificity expression of napsin A in adenocarcinomas of lung is useful to distinguish primary lung adenocarcinomas from adenocarcinomas of other organs. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title