Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Napsin-A Mouse anti-Human, Clone: NAPSA/1238 + NAPSA/1239- Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

$378.35

Specifications

Antigen Napsin A
Clone NAPSA/1238 + NAPSA/1239
Host Species Mouse
Isotype IgG Kappa
Gene Alias Asp 4, ASP4, Aspartyl protease 4, EC 3.4.23, EC 3.4.23.-, EC 3.4.23.15, EC 3.4.23.3, EC 3.4.23.5, KAP, Kdap, NAP1napsin-A, NAPApronapsin A, NAPSA, napsin A aspartic peptidase, napsin-1, SNAPA, TA01/TA02
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB254391100 View Documents Novus Biologicals
NBP254391100UG
100 ug Each for $378.35
Add to cart
 
Description

Description

Napsin A Monoclonal antibody specifically detects Napsin A in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications

Specifications

Napsin A
Mouse
Asp 4, ASP4, Aspartyl protease 4, EC 3.4.23, EC 3.4.23.-, EC 3.4.23.15, EC 3.4.23.3, EC 3.4.23.5, KAP, Kdap, NAP1napsin-A, NAPApronapsin A, NAPSA, napsin A aspartic peptidase, napsin-1, SNAPA, TA01/TA02
CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Store at -20 to -80C. Avoid freeze-thaw cycles.
Monoclonal
1 mg/mL
NAPSA/1238 + NAPSA/1239
IgG Kappa
Human
RUO
Purified
Primary
PBS with No Preservative

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit