Learn More
Napsin A Mouse anti-Human, Rat, DyLight 680, Clone: 10C4B8, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237355FR
Catalog No. NBP237355FR
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Napsin A
10C4B8
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
NAPSA
Purified recombinant fragment of human NAPSA (AA: 20-158) expressed in E. Coli.
Protein A or G purified
Cancer, Cell Biology, Cellular Markers
Primary
9476
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
Asp 4, ASP4, Aspartyl protease 4, EC 3.4.23, EC 3.4.23.-, EC 3.4.23.15, EC 3.4.23.3, EC 3.4.23.5, KAP, Kdap, NAP1napsin-A, NAPApronapsin A, NAPSA, napsin A aspartic peptidase, napsin-1, SNAPA, TA01/TA02
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Rat