Learn More
Inter Dyne Systems Inc NAR-LOCK
Supplier:  Inter Dyne Systems Inc NL2KD
Encompass_Preferred
stainless steel double locking narcotic cabinet w one removable interior shelf 21 in. l x 13 in. h x 10 in. deep. Unit is meant to be installed into an existing cabinet for security purposes.
Catalog No. NC9026391