Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NARC-1 (PCSK9) Mouse anti-Human, Unconjugated, Clone: 15A6, MilliporeSigma™

Mouse Monoclonal Antibody

$153.50 - $386.00

Specifications

Antigen NARC-1 (PCSK9)
Applications ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABS21611MI
View Documents
MilliporeSigma
MABS2161100UG
100 μg
Each for $386.00
Add to cart
 
MABS21612MI
View Documents
MilliporeSigma
MABS216125UG
25 μg
Each for $153.50
Add to cart
 
Description

Description

NARC-1 (PCSK9) Monoclonal antibody specifically detects NARC-1 (PCSK9) in Human samples. It is validated for ELISA, Immunocytochemistry, Immunoprecipitation, Western Blotting.
Specifications

Specifications

NARC-1 (PCSK9)
Unconjugated
Mouse
Proprotein convertase subtilisin/kexin type 9;Neural apoptosis-regulated convertase 1;Proprotein convertase 9;PC9;Subtilisin/kexin-like protease PC9.
Full-length purified human Proprotein convertase 9 (PCSK9).
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot
Purified
Human
PCSK9, NARC1, PSEC0052
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit