Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nardilysin (NRDC) Mouse anti-Human, Unconjugated, Clone: 102, MilliporeSigma™

Mouse Monoclonal Antibody

$153.50 - $386.00

Specifications

Antigen Nardilysin (NRDC)
Applications Immunoprecipitation, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABS20561MI
View Documents
MilliporeSigma
MABS2056100UL
100 μL
Each for $386.00
Add to cart
 
MABS20562MI
View Documents
MilliporeSigma
MABS205625UL
25 μL
Each for $153.50
Add to cart
 
Description

Description

Nardilysin (NRDC) Monoclonal antibody specifically detects Nardilysin (NRDC) in Human samples. It is validated for Immunoprecipitation, Western Blotting.
Specifications

Specifications

Nardilysin (NRDC)
Unconjugated
Mouse
N-arginine dibasic convertase;NRD convertase;NRDC;NRD-C;Nardilysin convertase
GST-tagged recombinant fragment corresponding to 80 amino acids from the N-terminal of human Nardilysin.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Immunoprecipitation, Western Blot
Purified
Human
NRDC, NRD1
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit