Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nardilysin (NRDC) Rat anti-Human,Mouse, Unconjugated, Clone: 135, MilliporeSigma™

Rat Monoclonal Antibody

$153.50 - $386.00

Specifications

Antigen Nardilysin (NRDC)
Applications ChIP assay, Immunocytochemistry, Immunoprecipitation, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rat
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABS20571MI
View Documents
MilliporeSigma
MABS2057100UG
100 μg
Each for $386.00
Add to cart
 
MABS20572MI
View Documents
MilliporeSigma
MABS205725UG
25 μg
Each for $153.50
Add to cart
 
Description

Description

Nardilysin (NRDC) Monoclonal antibody specifically detects Nardilysin (NRDC) in Human,Mouse samples. It is validated for Chromatin Immunoprecipitation (ChIP), Immunocytochemistry, Immunoprecipitation, Western Blotting.
Specifications

Specifications

Nardilysin (NRDC)
Unconjugated
Rat
N-arginine dibasic convertase;NRD convertase;NRDC;NRD-C;Nardilysin convertase
Full-length recombinant mouse Nardilysin expressed in silkworm expression system.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
ChIP assay, Immunocytochemistry, Immunoprecipitation, Western Blot
Purified
Human, Mouse
Nrdc, Nrd1
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit