Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NARG2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen NARG2
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB095626
View Documents
Novus Biologicals
NBP303785100UL
100 μg
Each for $320.71
Add to cart
 
NB095627
View Documents
Novus Biologicals
NBP30378520UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

NARG2 Polyclonal antibody specifically detects NARG2 in Human samples. It is validated for Western Blot
Specifications

Specifications

NARG2
Unconjugated
Rabbit
Human
BRCC1FLJ11896, breast cancer cell 1, NMDA receptor regulated 2, NMDA receptor-regulated gene 2, NMDA receptor-regulated protein 2
Recombinant fusion protein containing a sequence corresponding to amino acids 660-800 of human NARG2 (NP_078887.2).
Polyclonal
Affinity purified
Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
79664
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit