Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
United Scientific™ Narrow Mouth Polypropylene Reagent Bottles
Click to view available options
Capacity:
1000 mL (33.8 oz.)
125 mL (4.23 oz.)
15 mL (0.51 oz.)
250 mL (8.45 oz.)
30 mL (1.01 oz.)
500 mL (16.9 oz.)
60 mL (2.03 oz.)
Description
- Autoclavable
- Narrow mouth
- Material: Polypropylene
- Include polypropylene caps
Specifications
Height (English) | 2.3 in. |
Through the Closure Dispensing | No |
Product Type | Reagent Bottle |
Capacity | 15 mL (0.51 oz.) |
Closure Material | Polypropylene |
Closure Size | 20 mm |
Height (Metric) | 58 mm |
Capacity (Metric) | 15 mL |
Capacity (English) | 0.51 oz. |
Color | Translucent |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction