Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NARS Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548120

Catalog No. PA548120

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine, monkey and mouse based on sequence homology.

Aminoacyl-tRNA synthetases are a class of enzymes that charge tRNAs with their cognate amino acids. Asparaginyl-tRNA synthetase is localized to the cytoplasm and belongs to the class II family of tRNA synthetases. The N-terminal domain represents the signature sequence for the eukaryotic asparaginyl-tRNA synthetases. [provided by RefSeq, Jul 2008]
Specifications

Specifications

NARS
Polyclonal
PBS with 0.09% sodium azide
O43776
Asparagine--tRNA ligase, cytoplasmic, Asparaginyl-tRNA synthetase, AsnRS, NARS
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4677
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
NARS
Liquid
NARS
KLH conjugated synthetic peptide between 24-53 amino acids from the N-terminal region of human NARS.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit