Learn More
Sklar InstrumentsSupplier Diversity Partner Econo Jansen Nasal Bayonet Forceps, Sklar, 6-1/2 in., Blunt, Straight, Serrated, Stainless Steel. PK/12. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 21605
Econo Jansen Nasal Bayonet Forceps, Sklar, 6-1/2 in., Blunt, Straight, Serrated, Stainless Steel. Sklar Jansen Bayonet thumb forceps are commonly used in nasal procedures for placing packing. The bayonet style handle and long jaws allow an unobstructed view when working in the nasal cavity. Serrated tips securely hold surgical materials. These forceps may also be used in intracranial procedures. Econo instruments are Sklar's floor-grade, disposable instruments. Made in Pakistan, they offer the greatest utility at the lowest cost. Econo instruments eliminate the need for cleaning and reprocessing. Because they are disposable, Econo instruments also reduce the likelihood of cross-contamination. Econo instruments are available in bulk quantities.
Catalog No. 50-143-7670