Learn More
Nasco Color Me Whirl-Pak™ Bags
Write-On Whirl-Pak™ bags make sample identification easy!
Manufacturer:  Nasco B01297RT
 View more versions of this product
Catalog No. 01-812-208
Add to cart
Description
Description
  • Write on strip
  • Tape and wire opening
  • Tyvek tabs
  • Sterilized
Specifications
Specifications
Isolating samples according to the colored tape
Sterile
0.076 mm
24 oz.
Red
9 in.
3 mil
6 in.
710 mL
23 cm