Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nasco Hydra Culture Kit Products for Science Education

For 30 students to maintain their own cultures of Hydra.

$56.50 - $69.70

Specifications

Quantity Each
For Use With (Application) For 30 students to maintain their own hydra cultures.
Sufficient For Class of 30 students
View More Specs

Includes: Kit includes a culture of living Hydra (LM00029M); brine shrimp hatchery complete with eggs and salt; one unit of Invertebrate Culture Solution, which makes 5 liters of media and saves you extra shipping costs because it is shipped in concentrate; 1-gal. plastic aquarium and a pipette

Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
S06614 Products for Science Education
View Documents
Nasco
LM00674
Hydra Culture Kit
Each for $56.50
Add to cart
 
S06614C Products for Science Education
View Documents
Nasco
LC00674
Hydra culture kit; Live material coupon
Each for $69.70
Add to cart
 
Description

Description

Specifications

Specifications

Each
For 30 students to maintain their own hydra cultures.
Class of 30 students
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit