Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Nasco Whirl-Pak™ Pipette Filter Sleeve
Prevents solids from plugging the tip of the pipette when sampling liquids that may contain solid particles.
Manufacturer:  Nasco B01523WA
Catalog No. 01-812-331
Description
Description
  • 330 micron pores
  • Holds a 10 mL pipette
Specifications
Specifications
Pipette Filter Sleeve
3 mil
Polyethylene
25/PK.