Learn More
Andwin Scientific
Yamato 13.75L Natural Convection Oven, Programmable, High Temperature, 115V/220V
Manufacturer: Andwin Scientific DR200
Programmable 13.75L natural convection oven with high accuracy control at high temperature range from 300°C to 700°C. Equipped with high operability temperature controller. Can be used as constant temperature oven, drying oven, and electric furnace for ashing, sintering, etc. as unit is suitable for each of these three functions. Temperature, measured temperature and overheat prevention temperature can be digitally set by operation menu and ▲/▼ keys. Easy programmable operation, fixed temperature, quick auto stop, auto stop and auto start.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.