Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA595422
Description
Synthetic peptide sequence: 172-201aa, NEEFLHSDLLEDSKYRKIYSFTLKPRTIED. Reconstitution information: Add 0.2 mL of distilled water will yield a concentration of 500 μg/mL.
Arylamine N-acetyltransferases (NAT-1 and NAT-2) catalyze N- or O-acetylation of heterocyclic and arylamine substrates in the detoxification of a wide array of drugs. Certain alleles causing high N-acetyltransferase activity have been associated with colon and urinary bladder cancers, as NATs also bioactivate several known carcinogens. Both NAT-1 and NAT-2 are cytoplasmic proteins and play an active role in the detoxification of many arylamine and hydrazine drugs. N-acetylation polymorphism is determined by the level of NAT activity in liver tissues, and has been linked to the action and toxicity of drugs that contain amines. Human NAT-1 is the functional homolog of rodent NAT-2, while human NAT-2 is the functional homolog of rodent NAT-1.Specifications
NAT1 | |
Polyclonal | |
0.2mg sodium phosphate with 0.9MG NaCl, 0.9MG NaCl and 0.05MG sodium azide, 0.05MG sodium azide | |
P18440 | |
NAT1 | |
A synthetic peptide corresponding to a sequence at the C-terminus of human NAT1, different from the related mouse sequence by six amino acids, and from the related rat sequence by five amino acids. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
NAT1 | |
Arylamine N-acetyltransferase 1; 2.3.1.5; Arylamide acetylase 1; Monomorphic arylamine N-acetyltransferase; MNAT; N-acetyltransferase type 1; NAT-1; NAT1; AAC1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
9 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title