Learn More
NAT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 191881AP
Catalog No. 50-173-2364
Add to cart
Description
Description
Arylamine N-acetyltransferases (NAT-1 and NAT-2) catalyze N- or O-acetylation of heterocyclic and arylamine substrates in the detoxification of a wide array of drugs. Certain alleles causing high N-acetyltransferase activity have been associated with colon and urinary bladder cancers, as NATs also bioactivate several known carcinogens. Both NAT-1 and NAT-2 are cytoplasmic proteins and play an active role in the detoxification of many arylamine and hydrazine drugs. N-acetylation polymorphism is determined by the level of NAT activity in liver tissues, and has been linked to the action and toxicity of drugs that contain amines. Human NAT-1 is the functional homolog of rodent NAT-2, while human NAT-2 is the functional homolog of rodent NAT-1.
Specifications
Specifications
NAT1
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P18440, P50294, P50297
AAC1, Arylamide acetylase 1, MNAT, N acetyltransferase type 1, NAT 1, NAT1, NATI
Rabbit
IgG
150 μL
-20°C
Primary
116631, 17960, 9
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
NAT1
Liquid
Nat1
NAT1 Fusion Protein Ag6280
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat