Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAA30 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548142

Catalog No. PA548142

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Catalytic subunit of the N-terminal acetyltransferase C (NatC) complex. Catalyzes acetylation of the N-terminal methionine residues of peptides beginning with Met-Leu-Ala and Met-Leu-Gly. Necessary for the lysosomal localization and function of ARL8B sugeesting that ARL8B is a NatC substrate.
Specifications

Specifications

NAA30
Polyclonal
PBS with 0.09% sodium azide
Q147X3, Q8CES0
N-alpha-acetyltransferase 30, N-acetyltransferase 12, N-acetyltransferase MAK3 homolog, NatC catalytic subunit, NAA30, C14orf35, MAK3, NAT12
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
122830, 70646
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
NAA30
Liquid
NAA30
KLH conjugated synthetic peptide between 315-343 amino acids from the C-terminal region of human NAT12.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit