Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAT2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549606

Catalog No. PA549606

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Participates in the detoxification of a plethora of hydrazine and arylamine drugs. Catalyzes the N- or O-acetylation of various arylamine and heterocyclic amine substrates and is able to bioactivate several known carcinogens.
Specifications

Specifications

NAT2
Polyclonal
PBS with 0.09% sodium azide
P11245, P50295
Arylamine N-acetyltransferase 2, Arylamide acetylase 2, N-acetyltransferase type 2, NAT-2, Polymorphic arylamine N-acetyltransferase, PNAT, NAT2, AAC2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10, 17961
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
NAT2
Liquid
NAT2
KLH conjugated synthetic peptide between 226-254 amino acids from the C-terminal region of human NAT2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit