Learn More
Aviva Systems Biology Corp NAA80 Antibody - C-terminal region (ARP48733_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP48733_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NAT6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human NAT6. Alternate names: FUS-2, FUS2ApplicationWB. Species reactivity: Human, Yeast
Catalog No. 50-014-59905