Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAT8 Mouse anti-Human, DyLight™ 680, Clone: OTI5A8, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272878FR
Description
NAT8 Monoclonal specifically detects NAT8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
NAT8 | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
NAT8 | |
Full-length protein expressed in 293T cell transfected with human NAT8 expression vector. | |
0.1 mL | |
9027.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI5A8 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Camello-like protein 1, CML1, EC 2.3.1, EC 2.3.1.-, GLAkidney- and liver-specific gene product, Hcml1, N-acetyltransferase 8, N-acetyltransferase 8 (GCN5-related, putative), probable N-acetyltransferase 8, TSC501N-acetyltransferase 8 (camello like), TSC510 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title