Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAT8 Monoclonal Antibody (OTI6H3), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA524948
Description
NAT8 Monoclonal antibody specifically detects NAT8 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
NAT8 | |
Monoclonal | |
1.1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9UHE5 | |
NAT8 | |
Full-length protein expressed in 293T cell transfected with human NAT8 expression vector | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
OTI6H3 | |
Unconjugated | |
NAT8 | |
0610037O16Rik; Acetyltransferase 2; ATase2; camello-like 4; camello-like protein 1; Camello-like protein 4; CCNAT; CML1; Cml4; Cysteinyl-conjugate N-acetyltransferase; GLA; Hcml1; kidney- and liver-specific gene product; N-acetyltransferase 8; N-acetyltransferase 8 (camello like); N-acetyltransferase 8 (GCN5-related, putative); N-acetyltransferase 8 (putative); NAT8; probable N-acetyltransferase 8; probable N-acetyltransferase CML4; putative N-acetyltransferase Camello 4; TSC501; TSC510 | |
Mouse | |
Affinity Chromatography | |
RUO | |
9027 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction