Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAT8L Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549536

Catalog No. PA549536


Add to cart

Description

Description

Predicted to react with mouse and rat based on sequence homology.

Plays a role in the regulation of lipogenesis by producing N-acetylaspartate acid (NAA), a brain-specific metabolite. NAA occurs in high concentration in brain and its hydrolysis plays a significant part in the maintece of intact white matter. Promotes dopamine uptake by regulating TNF-alpha expression. Attenuates methamphetamine-induced inhibition of dopamine uptake.
Specifications

Specifications

NAT8L
Polyclonal
Unconjugated
NAT8L
Liquid
NAT8L
KLH conjugated synthetic peptide between 59-93 amino acids from human NAT8L.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q8N9F0, Q3UGX3
N-acetylaspartate synthetase, NAA synthetase, 2.3.1.17, Camello-like protein 3, N-acetyltransferase 8-like protein, NAT8L, CML3
Rabbit
IgG
50 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
269642, 339983
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit