Learn More
Selleck Chemical LLCSupplier Diversity Partner Nateglinide 10mg 105816-04-4 A-4166  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S248910MG
Nateglinide (A-4166) is an insulin secretagog agent that lowers blood glucose levels by stimulating insulin secretion from the pancreas.
Catalog No. 50-617-9