Learn More
Thermo Scientific™ National 12 x 96mm Glass Vials for Automated TOC analyzers Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ National 12 x 96mm Glass Vials for Automated TOC Analyzers are available in open top or screw-thread designs.
Manufacturer:  Thermo Scientific™ C40101296
 View more versions of this product
Catalog No. 03-397-12
  Check Availability
Add to cart
Description
Description
Thermo Scientific™ National 12 x 96mm Glass Vials for Automated TOC Analyzers are available in open top or screw-thread designs.
  • Maximum fill volume: 5mL
  • Superior quality N-51A clear borosilicate glass
  • 5mL sample vial is designed for use in the Shimadzu™ Model ASI-5000
  • Screw thread vial fits standard 10-425 closures (included with pre-assembled product)
  • Open-top vial was designed in cooperation with the original equipment manufacturer
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Shimadzu™ Model ASI-5000
10-425mm
Screw Thread TOC Vial, Polyprop. Cap
Screw Thread TOC Vial
Screw Thread TOC Vial, Polypropylene Cap with PTFE septum
5mL