Learn More
Thermo Scientific™ National 20mm Crimp Seals with Septa for Headspace Vials
Minimize the risk of contamination with quick, convenient Thermo Scientific™ National 20mm Crimp Seals with Septa.
Manufacturer:  Thermo Scientific™ C402031
 View more versions of this product
Catalog No. 03-397-45
  Check Availability
Add to cart
Description
Description
Minimize the risk of contamination with quick, convenient Thermo Scientific™ National 20mm Crimp Seals with Septa.
20mm snap cap with prefitted septa for headspace vials
  • Seals with pre-inserted septa are convenient and minimize contamination due to handling
  • Use magnetic seals with CTC/Leap™ Technologies, Gerstel™ and other magnetic transport autosamplers
  • PurePak™ PTFE/Silicone seals are packed in glass jars to maintain purity up to the point of use
  • Molded polyethylene SepCap™ is suitable for short-term storage or where repeated access to the vial is required
Ordering Information:
Septa and snap cap also sold separately.
Note:
SepCap is for short-term storage. Do not use SepCap at elevated temperatures.
Specifications
Specifications
Molded Polyethylene
SepCap Integral molded Polyethylene septa
SepCap Integral molded Polyethylene
C4020-31
SepCap Integral molded Polyethylene septa
20mm
headspace vials
National™