Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ National Assembled Certified Vial Kits

Thermo Scientific™ National Assembled Certified Vial Kits provide convenience and assurance vials uphold rigorous manufacturing standards.

Manufacturer:  Thermo Scientific™ CERT401588

 View more versions of this product

Catalog No. 03-377-502

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Thermo Scientific™ National Assembled Certified Vial Kits provide convenience and assurance vials uphold rigorous manufacturing standards.

  • Fully lot tested including HPLC and GC analysis
  • Certificate of Conformance included in every pack
  • Bonded caps to prevent septa push-through
  • Clear Type 1, 33 expansion borosilicate glass or amber Type 2, 51 expansion borosilicate glass
  • Available with or without ID patch
  • Fixed micro vials or inserts for limited-volume sampling
  • Assembled in convenient packs of 100 vials and pre-attached caps and septa
Specifications

Specifications

limited-volume sampling
13mm
Black
Screw Thread
2mL Clear Glass Vial Kit, 13mm Screw Thread, PTFE/Silicone Closure
Clear
Assembled Vial Kit
2mL
PTFE/Silicone Septum
2mL Clear Vial Kit, 13mm Screw Thread
Red PTFE/White Silicone
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit