Thermo Scientific National Assembled Convenience Kits: EPA Vials with Caps and Septa

Thermo Scientific™ National EPA Vial Convenience Kits are designed for discrete water sampling applications.

Manufacturer: Thermo Scientific B7920C

 View more versions of this product

Catalog No. 03-377-33

  Check Availability
Add to Cart

    Specifications

    For Use With discrete water sampling applications
    Item Description B7920-C
    Certifications/Compliance Class 200
    Type Assembled EPA Vial Kit
    Volume (Metric) 20mL
    Color Clear
    Description Clear; 20mL

    Thermo Scientific™ National EPA Vial Convenience Kits are designed for discrete water sampling applications.

    • Recommended for discrete water sampling under EPA 40 CFR 136 “Guidelines for Establishing Test Procedures for the Analysis of Pollutants” and EPA 40 CFR 141 “National Interim Primary Drinking Water Regulations: Control of Trihalomethanes in Drinking Water”
    • Class 100—Vials, Caps and Septa are assembled
    • Class 200—Vials, Caps and Septa are processed to EPA specifications and assembled
    • Class 300—Vials, Caps and Septa are processed and certified to EPA specifications, then assembled

    Includes:

    Certificate of Analysis is included with each box of vials.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit