Learn More
Thermo Scientific™ National Assembled Convenience Kits: EPA Vials with Caps and Septa Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ National EPA Vial Convenience Kits are designed for discrete water sampling applications.
Manufacturer:  Thermo Scientific™ B7950C3
 View more versions of this product
Catalog No. 03-377-303
  Check Availability
Add to cart
Specifications
Description
Thermo Scientific™ National EPA Vial Convenience Kits are designed for discrete water sampling applications.
  • Recommended for discrete water sampling under EPA 40 CFR 136 “Guidelines for Establishing Test Procedures for the Analysis of Pollutants” and EPA 40 CFR 141 “National Interim Primary Drinking Water Regulations: Control of Trihalomethanes in Drinking Water”
  • Class 100—Vials, Caps and Septa are assembled
  • Class 200—Vials, Caps and Septa are processed to EPA specifications and assembled
  • Class 300—Vials, Caps and Septa are processed and certified to EPA specifications, then assembled
Includes:
Certificate of Analysis is included with each box of vials.
Description & Specifications
Specifications
discrete water sampling applications
40mL
Clear; 40mL
Class 300
Assembled EPA Vial Kit
Clear
B7950-C3