Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits provide convenience, efficiency and stackable storage options.
Manufacturer: Thermo Scientific™ C401357A
 View more versions of this product
Catalog No. 03-375-50C
  Check Availability
Add to Cart
    Specifications
    Vial Volume (Metric) 2mL
    Septa Red PTFE/White Silicone
    Color Amber
    Type Assembled Vial Kit
    Vial Closure Size 8-425mm
    Closure Color Black
    Closure Material Polypropylene
    Closure Type Polypropylene, Flanged
    Item Description C4013-57A
    Description Amber Glass; PP Black Cap; Red PTFE/White Silicone septum; in storage tray
    Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits provide convenience, efficiency and stackable storage options.
    • Packaged in convenient vial trays with clear covers or in economical polybags
    • Vial trays are stackable for easy storage
    • All caps are manufactured from virgin polypropylene unless noted
    Includes: 
    100 vials with pre-attached caps and septa
  • Description & Specifications
Related Products
Product Content Feedback