Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits provide convenience, efficiency and stackable storage options.
Manufacturer: Thermo Scientific™ C4013016A
 View more versions of this product
Catalog No. 03-375-38E
  Check Availability
Add to Cart
    Specifications
    Septa Red PTFE/White Silicone/Red PTFE
    Type Assembled Vial Kit
    Closure Color Black
    Closure Type Polypropylene
    Color Clear
    Vial Closure Size 8-425mm
    Vial Volume (Metric) 2mL
    Closure Material Polypropylene
    Item Description C4013-016A
    Description Clear Glass; Black cap; Red PTFE/White Silicone/Red PTFE septum; in polybag
    Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits provide convenience, efficiency and stackable storage options.
    • Packaged in convenient vial trays with clear covers or in economical polybags
    • Vial trays are stackable for easy storage
    • All caps are manufactured from virgin polypropylene unless noted
    Includes: 
    100 vials with pre-attached caps and septa
  • Description & Specifications
Related Products
Product Content Feedback