Learn More
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits provide convenience, efficiency and stackable storage options.
Manufacturer:  Thermo Scientific™ C401319A
 View more versions of this product
Catalog No. 03-375-20A
  Check Availability
Add to cart
Description
Description
Thermo Scientific™ National Assembled Standard Opening Vial, Cap and Septum Kits provide convenience, efficiency and stackable storage options.
  • Packaged in convenient vial trays with clear covers or in economical polybags
  • Vial trays are stackable for easy storage
  • All caps are manufactured from virgin polypropylene unless noted
Includes: 
100 vials with pre-attached caps and septa
Specifications
Specifications
Assembled Vial Kit
2mL
Polypropylene
C4013-19A
White Virgin PTFE
8-425mm
Black
Polypropylene
Amber Glass; PP Black Cap; White Virgin PTFE septum; in storage tray
Amber