Learn More
National Diagnostics™ Calci-Clear™ Decalcifying Solution
A solution of efficient chelating and sequestering agents used with tissue samples to remove and bind all calcium and other metal ions
$50.40 - $485.90
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-899-90139
|
National Diagnostics™
HS1041QRT |
1 qt. |
Each for $50.40
|
|
|||||
50-899-90138
|
National Diagnostics™
HS1041GLL |
1 gal. |
Each for $116.82
|
|
|||||
50-899-90140
|
National Diagnostics™
HS1045GLL |
5 gal. |
Each for $485.90
|
|
|||||
Description
- Moderate speed decalcifier
- Mildness balanced with performance
- Reduces interference with subsequent staining
- Minimizes tissue distortion and structure loss
- May be used to decalcify any tissue
Specifications
Decalcifying Solution |