Learn More
Thermo Scientific™ National Inserts for 15 x 45mm 4mL Vials Available on GSA/VA Contract for Federal Government customers only.
Thermo Scientific™ National Vial Inserts offer a variety of options incuding glass conical, Polyspring and pulled point.
Manufacturer:  Thermo Scientific™ C4015S642
 View more versions of this product
Catalog No. 03-377-434
  Check Availability
Add to cart
Description
Description
Thermo Scientific™ National Vial Inserts offer a variety of options incuding glass conical, Polyspring and pulled point.
  • Have a nominal O.D. of 6 or 8mm to fit 15 x 45 4mL vials
  • Deactivated inserts are NSC Silanized
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
15 x 45mm 4mL Vials
Vial Insert
C4015-S642
National™
Silanized Conical Insert, Pulled Point
350µL