Learn More
Thermo Scientific™ National Inserts for Standard-Opening Vials
With a nominal O.D. of 5mm, these Thermo Scientific™ National Inserts fit most standard-opening crimp top vials.
$152.00
Specifications
For Use With Standard-Opening Vials
View More Specs
Products 1
Catalog Number Mfr. No. Type Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Type Volume (Metric) Price Quantity & Availability  
03-378-359
View Documents
Thermo Scientific™
C4012K530
Vial Insert 200 μL
Pack of 100 for $152.00
Add to cart
Description
Description
With a nominal O.D. of 5mm, these Thermo Scientific™ National Inserts fit most standard-opening crimp top vials.
  • Pulled point inserts are an economical choice for non-critical applications
Specifications
Specifications
Standard-Opening Vials