National Marker™ Biohazard: Authorized Personnel Only Sign

Sign for hazardous areas are emphasized by graphic

Manufacturer: National Marker Co.Supplier Diversity Partner SA165R

 View more versions of this product

Catalog No. 18-999-756

  Check Availability
Add to Cart

    Specifications

    Message Biohazard Authorized Personnel Only
    Quantity Each
    Length (English) 7 in.
    Material Plastic
    Certifications/Compliance OSHA
    Type Safety sign
    Width (English) 17 in.,17 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit