Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
National Marker™ Biohazard: Caution Biological Hazard Sign
Emphasizes safety message with the use of a biohazard graphic
Specifications
Certifications/Compliance | OSHA |
---|---|
Quantity | 1 Ea. |
Height (English) | 7 in. |
Width (English) | 17 in. |
Displayed Text | BIOHAZARD CAUTION BIOLOGICAL HAZARD/Biohazard Pictogram |
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Price | Quantity & Availability | |||||
18-999-758
|
National Marker™
SA52P |
Vinyl | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
17-984-10A
|
National Marker™
SA 52 R |
Plastic | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Orange sign with black text and graphic
- Available in rigid plastic or pressure-sensitive vinyl
- Size: 17.7 × 43.2cm (7 in. × 17 in.)
Specifications
OSHA | |
7 in. | |
BIOHAZARD CAUTION BIOLOGICAL HAZARD/Biohazard Pictogram |
1 Ea. | |
17 in. | |
Biohazard Sign |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title