National Marker™ Chemical Hazard Identification Labels

High visibility red-and-white pressure-sensitive vinyl labels ideal for alerting employees to potential chemical hazards in the workplace

Manufacturer: National Marker Co.Supplier Diversity Partner HC281P

 View more versions of this product

Catalog No. 17-985-464

  Check Availability
Add to Cart

    Specifications

    Display Text Nitric Acid
    Format Label
    Height (English) 6 in.
    For Use With (Application) For marking larger containers, both indoor and outdoor applications
    Height (Metric) 15.24cm
    Material Vinyl
    Product Type Container Label
    Quantity Each
    Color Red on White
    Width (English) 10 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit