Learn More
National Marker™ Colored Delineator Tape
Clear laminate on tape surface resists abrasion, dirt, oil, and chemicals
Manufacturer:  National Marker Co.Supplier Diversity Partner T207 18
 View more versions of this product
Catalog No. 17-985-484A
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
54.13 ft. x 2 in. (16.5m x 5.08cm)
Magenta,Yellow
Delineator Tape
Floor markings in high traffic areas
6mil
5.08cm
OSHA 1910.144
Vinyl
54.13 ft.
16.5m
2 in.
Pressure sensitive