Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Danger Unsafe Do Not Use Tags
Warn of potential hazards and provide information on the job site
Includes: Self-locking nylon fasteners
Description
- Material: Unrippable vinyl
- Size: 15.24 × 7.62cm (6 × 3 in.)
Specifications
Vinyl | |
15.2 cm | |
3 in. | |
6 x 3 in. (15.2 x 7.62 cm) |
DANGER Unsafe Do Not Use | |
6 in. | |
7.62 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title