National Marker DOT Shipping Labels

For marking packages with hazardous materials

 View more versions of this product

Catalog No. 17-987-171A

  Check Availability
Add to Cart

    Specifications

    Display Text Corrosive
    Product Type DOT Shipping Label
    Quantity 500 Roll(s)
    Color Black, White
    Width (English) 4 in.
    Format Label
    Height (English) 4 in.
    For Use With (Application) Designed specifically for vehicles dedicated to the repeat transportation of a single material
    Height (Metric) 10.16cm
    Material Paper
    Certifications/Compliance DOT 49 CFR 172.519

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit