National Marker Glo-Brite Exit Signs

Manufacturer: National Marker Co.Supplier Diversity Partner 7010B

 View more versions of this product

Catalog No. 19-065-996

  Check Availability
Add to Cart

    Specifications

    Includes Hardware
    Quantity Each
    Style Wall Mount
    Color Red,Red
    Type Bracket
    Width (English) 4 in.,4 in.
    For Use With (Application) For Glo Brite™ exit sign
    Height (English) 4 in.,4 in.
    Material Plastic

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit