Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Marker™ “Lock-Out Center” Sign

$23.55 - $43.00

Specifications

Message Lock Out Center
Quantity 1 Ea.
Height (English) 7 in.
Type Safety Sign
Width (English) 17 in.
View More Specs

Products 2
Catalog Number Mfr. No. Material Price Quantity & Availability  
Catalog Number Mfr. No. Material Price Quantity & Availability  
18-999-746
View Documents
National Marker™
SA148P
Vinyl
Each for $23.55
Add to cart
 
18-999-745
View Documents
National Marker™
SA148R
Plastic
Each for $43.00
Add to cart
 
Description

Description

Specifications

Specifications

Lock Out Center
7 in.
17 in.
1 Ea.
Safety Sign
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit