National Marker Rigid Plastic Glow-in-the Dark Signs

Manufacturer: National Marker Co.Supplier Diversity Partner GL 16 R

 View more versions of this product

Catalog No. 18-999-919

  Check Availability
Add to Cart

    Specifications

    Height (English) 10 in.,10 in.
    Certifications/Compliance OSHA 1910.37, ASTM E2072-00, ASTM E2073-02, ASTM E2030-02, ASTM E162, ASTM E648, ASTM E662,OSHA 1910.37, ASTM E2072-00, ASTM E2073-02, ASTM E2030-02, ASTM E162, ASTM E648, ASTM E662
    Message Exit With Left Arrow
    Quantity Each
    Thickness 0.05 in.
    Type Glow-in-the dark sign
    For Use With (Application) Indoor and outdoor use
    Material Phosphorescent
    Style Rigid
    Width (English) 10 in.,10 in.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit